Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM35A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM35A - family with sequence similarity 35 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019054.2 | Intron | NP_061927.2 | ||||
XM_005269920.3 | Intron | XP_005269977.1 | ||||
XM_006717903.3 | Intron | XP_006717966.1 | ||||
XM_006717905.3 | Intron | XP_006717968.1 | ||||
XM_011539876.2 | Intron | XP_011538178.1 | ||||
XM_011539877.2 | Intron | XP_011538179.1 | ||||
XM_011539878.2 | Intron | XP_011538180.1 | ||||
XM_011539879.2 | Intron | XP_011538181.1 | ||||
XM_011539881.2 | Intron | XP_011538183.1 | ||||
XM_011539882.2 | Intron | XP_011538184.1 | ||||
XM_011539883.2 | Intron | XP_011538185.1 | ||||
XM_017016349.1 | Intron | XP_016871838.1 | ||||
XM_017016350.1 | Intron | XP_016871839.1 | ||||
XM_017016351.1 | Intron | XP_016871840.1 | ||||
XM_017016352.1 | Intron | XP_016871841.1 | ||||
XM_017016353.1 | Intron | XP_016871842.1 |