Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611908 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFT1 - RFT1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052859.3 | 5266 | Intron | NP_443091.1 | |||
XM_005265537.4 | 5266 | Intron | XP_005265594.1 | |||
XM_006713384.3 | 5266 | Intron | XP_006713447.1 | |||
XM_011534214.2 | 5266 | UTR 3 | XP_011532516.1 | |||
XM_011534215.2 | 5266 | UTR 3 | XP_011532517.1 | |||
XM_011534216.2 | 5266 | Intron | XP_011532518.1 | |||
XM_017007460.1 | 5266 | Intron | XP_016862949.1 | |||
XM_017007461.1 | 5266 | Intron | XP_016862950.1 |