Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602545 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTPRK - protein tyrosine phosphatase, receptor type K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135648.2 | Intron | NP_001129120.1 | ||||
NM_001291981.1 | Intron | NP_001278910.1 | ||||
NM_001291982.1 | Intron | NP_001278911.1 | ||||
NM_001291983.1 | Intron | NP_001278912.1 | ||||
NM_001291984.1 | Intron | NP_001278913.1 | ||||
NM_002844.3 | Intron | NP_002835.2 | ||||
XM_006715537.2 | Intron | XP_006715600.1 | ||||
XM_011536014.2 | Intron | XP_011534316.1 | ||||
XM_011536015.2 | Intron | XP_011534317.1 | ||||
XM_011536016.2 | Intron | XP_011534318.1 | ||||
XM_011536017.2 | Intron | XP_011534319.1 | ||||
XM_011536018.2 | Intron | XP_011534320.1 | ||||
XM_011536020.2 | Intron | XP_011534322.1 | ||||
XM_011536021.2 | Intron | XP_011534323.1 | ||||
XM_017011145.1 | Intron | XP_016866634.1 | ||||
XM_017011146.1 | Intron | XP_016866635.1 | ||||
XM_017011147.1 | Intron | XP_016866636.1 | ||||
XM_017011148.1 | Intron | XP_016866637.1 | ||||
XM_017011149.1 | Intron | XP_016866638.1 | ||||
XM_017011150.1 | Intron | XP_016866639.1 |
Set Membership: |
HapMap |