Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609380 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMLN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LMLN - leishmanolysin like peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136049.2 | 323 | Missense Mutation | GAG,GAT | E,D 106 | NP_001129521.2 | |
NM_033029.3 | 323 | Missense Mutation | GAG,GAT | E,D 106 | NP_149018.2 | |
XM_011513270.1 | 323 | Missense Mutation | GAG,GAT | E,D 106 | XP_011511572.1 | |
XM_011513271.1 | 323 | Missense Mutation | GAG,GAT | E,D 106 | XP_011511573.1 | |
XM_017007428.1 | 323 | Missense Mutation | GAG,GAT | E,D 106 | XP_016862917.1 |
Set Membership: |
HapMap |