Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615692 MIM: 602644 | ||||||||||||||||||||
Literature Links: |
CHID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHID1 - chitinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142674.1 | Intron | NP_001136146.1 | ||||
NM_001142675.1 | Intron | NP_001136147.1 | ||||
NM_001142676.1 | Intron | NP_001136148.1 | ||||
NM_001142677.1 | Intron | NP_001136149.1 | ||||
NM_023947.3 | Intron | NP_076436.3 | ||||
XM_005253072.2 | Intron | XP_005253129.1 | ||||
XM_005253073.4 | Intron | XP_005253130.1 | ||||
XM_006718282.2 | Intron | XP_006718345.1 | ||||
XM_011520299.1 | Intron | XP_011518601.1 | ||||
XM_011520300.1 | Intron | XP_011518602.1 | ||||
XM_011520301.2 | Intron | XP_011518603.1 | ||||
XM_011520302.2 | Intron | XP_011518604.1 | ||||
XM_017018166.1 | Intron | XP_016873655.1 | ||||
XM_017018167.1 | Intron | XP_016873656.1 | ||||
XM_017018168.1 | Intron | XP_016873657.1 |
TSPAN4 - tetraspanin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |