Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611871 MIM: 602307 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RMDN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WWP1 - WW domain containing E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007013.3 | Intron | NP_008944.1 | ||||
XM_005250760.3 | Intron | XP_005250817.1 | ||||
XM_005250761.3 | Intron | XP_005250818.1 | ||||
XM_011516802.2 | Intron | XP_011515104.1 | ||||
XM_017012993.1 | Intron | XP_016868482.1 | ||||
XM_017012994.1 | Intron | XP_016868483.1 | ||||
XM_017012995.1 | Intron | XP_016868484.1 | ||||
XM_017012996.1 | Intron | XP_016868485.1 | ||||
XM_017012997.1 | Intron | XP_016868486.1 |
Set Membership: |
HapMap |