Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604819 MIM: 607733 | ||||||||||||||||||||
Literature Links: |
MIR937 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR937 - microRNA 937 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PUF60 - poly(U) binding splicing factor 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136033.2 | 739 | Missense Mutation | GCT,GTT | A,V 176 | NP_001129505.1 | |
NM_001271096.1 | 739 | Missense Mutation | GCT,GTT | A,V 201 | NP_001258025.1 | |
NM_001271097.1 | 739 | Missense Mutation | GCT,GTT | A,V 173 | NP_001258026.1 | |
NM_001271098.1 | 739 | Missense Mutation | GCT,GTT | A,V 218 | NP_001258027.1 | |
NM_001271099.1 | 739 | Missense Mutation | GCT,GTT | A,V 190 | NP_001258028.1 | |
NM_001271100.1 | 739 | Missense Mutation | GCT,GTT | A,V 159 | NP_001258029.1 | |
NM_014281.4 | 739 | Missense Mutation | GCT,GTT | A,V 202 | NP_055096.2 | |
NM_078480.2 | 739 | Missense Mutation | GCT,GTT | A,V 219 | NP_510965.1 | |
XM_011516929.2 | 739 | Missense Mutation | GCT,GTT | A,V 256 | XP_011515231.1 | |
XM_011516930.2 | 739 | Missense Mutation | GCT,GTT | A,V 239 | XP_011515232.1 | |
XM_017013234.1 | 739 | Missense Mutation | GCT,GTT | A,V 256 | XP_016868723.1 | |
XM_017013235.1 | 739 | Missense Mutation | GCT,GTT | A,V 244 | XP_016868724.1 | |
XM_017013236.1 | 739 | Missense Mutation | GCT,GTT | A,V 239 | XP_016868725.1 | |
XM_017013237.1 | 739 | Missense Mutation | GCT,GTT | A,V 256 | XP_016868726.1 | |
XM_017013238.1 | 739 | Missense Mutation | GCT,GTT | A,V 256 | XP_016868727.1 | |
XM_017013239.1 | 739 | Missense Mutation | GCT,GTT | A,V 176 | XP_016868728.1 | |
XM_017013240.1 | 739 | Missense Mutation | GCT,GTT | A,V 159 | XP_016868729.1 |
SCRIB - scribbled planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |