Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGCCACCGTGGAGTCTTTAGTA[A/T]TTTGAAGGAAGTGTGATTAGATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100131655 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100131655 - uncharacterized LOC100131655 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF236 - zinc finger protein 236 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306089.1 | Intron | NP_001293018.1 | ||||
NM_007345.3 | Intron | NP_031371.3 | ||||
XM_005266762.3 | Intron | XP_005266819.1 | ||||
XM_011526165.2 | Intron | XP_011524467.1 | ||||
XM_011526166.2 | Intron | XP_011524468.1 | ||||
XM_011526168.2 | Intron | XP_011524470.1 | ||||
XM_011526169.2 | Intron | XP_011524471.1 |
Set Membership: |
HapMap |