Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613543 MIM: 610012 | |||||||||||||||||||||||
Literature Links: |
SLCO5A1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLCO5A1 - solute carrier organic anion transporter family member 5A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146008.1 | 5891 | Intron | NP_001139480.1 | |||
NM_001146009.1 | 5891 | Intron | NP_001139481.1 | |||
NM_030958.2 | 5891 | Intron | NP_112220.2 | |||
XM_005251313.2 | 5891 | UTR 3 | XP_005251370.1 | |||
XM_017013883.1 | 5891 | UTR 3 | XP_016869372.1 | |||
XM_017013884.1 | 5891 | Intron | XP_016869373.1 | |||
XM_017013885.1 | 5891 | UTR 3 | XP_016869374.1 | |||
XM_017013886.1 | 5891 | Intron | XP_016869375.1 |
SULF1 - sulfatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |