Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK1 - dedicator of cytokinesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290223.1 | Intron | NP_001277152.1 | ||||
NM_001380.4 | Intron | NP_001371.1 | ||||
XM_011539422.2 | Intron | XP_011537724.1 | ||||
XM_011539424.1 | Intron | XP_011537726.1 | ||||
XM_017015813.1 | Intron | XP_016871302.1 | ||||
XM_017015814.1 | Intron | XP_016871303.1 | ||||
XM_017015815.1 | Intron | XP_016871304.1 | ||||
XM_017015816.1 | Intron | XP_016871305.1 | ||||
XM_017015817.1 | Intron | XP_016871306.1 |