Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD4B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRMD4B - FERM domain containing 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015123.2 | 3384 | UTR 3 | NP_055938.1 | |||
XM_005264722.1 | 3384 | UTR 3 | XP_005264779.1 | |||
XM_005264723.1 | 3384 | UTR 3 | XP_005264780.1 | |||
XM_017005989.1 | 3384 | UTR 3 | XP_016861478.1 | |||
XM_017005990.1 | 3384 | UTR 3 | XP_016861479.1 | |||
XM_017005991.1 | 3384 | UTR 3 | XP_016861480.1 | |||
XM_017005992.1 | 3384 | UTR 3 | XP_016861481.1 | |||
XM_017005993.1 | 3384 | UTR 3 | XP_016861482.1 | |||
XM_017005994.1 | 3384 | UTR 3 | XP_016861483.1 | |||
XM_017005995.1 | 3384 | UTR 3 | XP_016861484.1 | |||
XM_017005996.1 | 3384 | UTR 3 | XP_016861485.1 | |||
XM_017005997.1 | 3384 | UTR 3 | XP_016861486.1 |