Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609804 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP52 - cilia and flagella associated protein 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080556.1 | Intron | NP_001074025.1 | ||||
NM_145054.4 | Intron | NP_659491.4 | ||||
XM_017024227.1 | Intron | XP_016879716.1 |
USP43 - ubiquitin specific peptidase 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267576.1 | Intron | NP_001254505.1 | ||||
NM_153210.4 | Intron | NP_694942.3 | ||||
XM_011523639.2 | Intron | XP_011521941.1 | ||||
XM_011523640.1 | Intron | XP_011521942.1 | ||||
XM_011523641.1 | Intron | XP_011521943.1 | ||||
XM_011523642.2 | Intron | XP_011521944.1 | ||||
XM_017024159.1 | Intron | XP_016879648.1 | ||||
XM_017024160.1 | Intron | XP_016879649.1 | ||||
XM_017024161.1 | Intron | XP_016879650.1 | ||||
XM_017024162.1 | Intron | XP_016879651.1 |
Set Membership: |
HapMap |