Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607262 MIM: 191318 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPN1 - epsin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130071.1 | Intron | NP_001123543.1 | ||||
NM_001130072.1 | Intron | NP_001123544.1 | ||||
NM_013333.3 | Intron | NP_037465.2 | ||||
XM_005258829.2 | Intron | XP_005258886.1 | ||||
XM_005258830.2 | Intron | XP_005258887.1 | ||||
XM_011526880.1 | Intron | XP_011525182.1 | ||||
XM_011526881.1 | Intron | XP_011525183.1 | ||||
XM_017026722.1 | Intron | XP_016882211.1 | ||||
XM_017026723.1 | Intron | XP_016882212.1 | ||||
XM_017026724.1 | Intron | XP_016882213.1 | ||||
XM_017026725.1 | Intron | XP_016882214.1 |
U2AF2 - U2 small nuclear RNA auxiliary factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |