Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607300 MIM: 607848 MIM: 607873 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRPF8 - pre-mRNA processing factor 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RILP - Rab interacting lysosomal protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031430.2 | Intron | NP_113618.2 | ||||
XM_005256811.1 | Intron | XP_005256868.1 | ||||
XM_017025195.1 | Intron | XP_016880684.1 |
SCARF1 - scavenger receptor class F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003693.3 | Intron | NP_003684.2 | ||||
NM_145350.2 | Intron | NP_663325.1 | ||||
XM_017025256.1 | Intron | XP_016880745.1 |
Set Membership: |
HapMap |