Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610874 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPATC1 - spermatogenesis and centriole associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134374.1 | Intron | NP_001127846.1 | ||||
NM_198572.2 | Intron | NP_940974.2 | ||||
XM_011517021.1 | Intron | XP_011515323.1 | ||||
XM_011517022.2 | Intron | XP_011515324.1 | ||||
XM_011517023.2 | Intron | XP_011515325.1 | ||||
XM_011517024.2 | Intron | XP_011515326.1 | ||||
XM_011517025.2 | Intron | XP_011515327.1 | ||||
XM_017013399.1 | Intron | XP_016868888.1 |