Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605337 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFHR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFHR4 - complement factor H related 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201550.2 | Intron | NP_001188479.1 | ||||
NM_001201551.1 | Intron | NP_001188480.1 | ||||
NM_006684.4 | Intron | NP_006675.2 | ||||
XM_006711129.3 | Intron | XP_006711192.1 | ||||
XM_017000110.1 | Intron | XP_016855599.1 | ||||
XM_017000111.1 | Intron | XP_016855600.1 | ||||
XM_017000112.1 | Intron | XP_016855601.1 | ||||
XM_017000113.1 | Intron | XP_016855602.1 | ||||
XM_017000114.1 | Intron | XP_016855603.1 |