Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142440 MIM: 600235 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HPN - hepsin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002151.2 | Intron | NP_002142.1 | ||||
NM_182983.2 | Intron | NP_892028.1 | ||||
XM_005258838.4 | Intron | XP_005258895.2 | ||||
XM_006723181.3 | Intron | XP_006723244.2 | ||||
XM_017026731.1 | Intron | XP_016882220.1 | ||||
XM_017026732.1 | Intron | XP_016882221.1 |
SCN1B - sodium voltage-gated channel beta subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |