Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTAATATTTCAACTTGAACAA[A/G]AAAACAAAGGCGTTAGAGAGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611667 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100335030 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100335030 - FGFR1 oncogene partner 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATS2 - spermatogenesis associated serine rich 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293285.1 | Intron | NP_001280214.1 | ||||
NM_001293286.1 | Intron | NP_001280215.1 | ||||
NM_023071.3 | Intron | NP_075559.2 | ||||
XM_011538690.2 | Intron | XP_011536992.1 | ||||
XM_011538691.2 | Intron | XP_011536993.1 | ||||
XM_017019840.1 | Intron | XP_016875329.1 |