Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606526 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLPH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MLPH - melanophilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042467.2 | Intron | NP_001035932.1 | ||||
NM_001281473.1 | Intron | NP_001268402.1 | ||||
NM_001281474.1 | Intron | NP_001268403.1 | ||||
NM_024101.6 | Intron | NP_077006.1 | ||||
XM_006712737.1 | Intron | XP_006712800.1 | ||||
XM_006712739.1 | Intron | XP_006712802.1 | ||||
XM_006712740.1 | Intron | XP_006712803.1 | ||||
XM_011511812.1 | Intron | XP_011510114.1 | ||||
XM_017004893.1 | Intron | XP_016860382.1 | ||||
XM_017004894.1 | Intron | XP_016860383.1 |
Set Membership: |
HapMap |