Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603625 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LY6H PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LY6H - lymphocyte antigen 6 complex, locus H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130478.1 | 596 | UTR 3 | NP_001123950.1 | |||
NM_001135655.1 | 596 | UTR 3 | NP_001129127.1 | |||
NM_002347.4 | 596 | UTR 3 | NP_002338.3 | |||
XM_017013413.1 | 596 | UTR 3 | XP_016868902.1 | |||
XM_017013414.1 | 596 | UTR 3 | XP_016868903.1 |