Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601750 MIM: 613846 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2H3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GTF2H3 - general transcription factor IIH subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCTN2 - tectonic family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143850.2 | 34 | UTR 5 | NP_001137322.1 | |||
NM_024809.4 | 34 | UTR 5 | NP_079085.2 | |||
XM_005253623.2 | 34 | UTR 5 | XP_005253680.1 | |||
XM_006719605.3 | 34 | UTR 5 | XP_006719668.1 | |||
XM_017019974.1 | 34 | UTR 5 | XP_016875463.1 | |||
XM_017019975.1 | 34 | UTR 5 | XP_016875464.1 |
Set Membership: |
HapMap |