Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608250 MIM: 616711 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUDS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SUDS3 - SDS3 homolog, SIN3A corepressor complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022491.2 | Intron | NP_071936.2 | ||||
XM_011538671.2 | Intron | XP_011536973.1 | ||||
XM_017019817.1 | Intron | XP_016875306.1 | ||||
XM_017019818.1 | Intron | XP_016875307.1 | ||||
XM_017019819.1 | Intron | XP_016875308.1 | ||||
XM_017019820.1 | Intron | XP_016875309.1 |
TAOK3 - TAO kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |