Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMN1 - antagonist of mitotic exit network 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113402.1 | Intron | NP_001106873.1 | ||||
NM_001278411.1 | Intron | NP_001265340.1 | ||||
NM_001278412.1 | Intron | NP_001265341.1 | ||||
XM_006719054.3 | Intron | XP_006719117.1 | ||||
XM_011520604.2 | Intron | XP_011518906.1 | ||||
XM_017018963.1 | Intron | XP_016874452.1 | ||||
XM_017018964.1 | Intron | XP_016874453.1 | ||||
XM_017018965.1 | Intron | XP_016874454.1 |
Set Membership: |
HapMap |