Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRAMD1A - GRAM domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136199.2 | Intron | NP_001129671.1 | ||||
NM_001320034.1 | Intron | NP_001306963.1 | ||||
NM_001320035.1 | Intron | NP_001306964.1 | ||||
NM_001320036.1 | Intron | NP_001306965.1 | ||||
NM_020895.4 | Intron | NP_065946.2 | ||||
XM_011527149.1 | Intron | XP_011525451.1 | ||||
XM_011527153.1 | Intron | XP_011525455.1 | ||||
XM_011527154.2 | Intron | XP_011525456.1 | ||||
XM_011527155.1 | Intron | XP_011525457.1 | ||||
XM_011527156.1 | Intron | XP_011525458.1 | ||||
XM_017027034.1 | Intron | XP_016882523.1 | ||||
XM_017027035.1 | Intron | XP_016882524.1 |