Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607811 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C6orf52 - chromosome 6 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145020.1 | Intron | NP_001138492.1 | ||||
XM_006715088.3 | Intron | XP_006715151.1 | ||||
XM_011514572.2 | Intron | XP_011512874.1 | ||||
XM_011514574.2 | Intron | XP_011512876.1 | ||||
XM_011514575.2 | Intron | XP_011512877.1 |
PAK1IP1 - PAK1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017906.2 | Intron | NP_060376.2 | ||||
XM_005249204.2 | Intron | XP_005249261.1 | ||||
XM_011514720.1 | Intron | XP_011513022.1 | ||||
XM_011514721.1 | Intron | XP_011513023.1 |
Set Membership: |
HapMap |