Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609360 | ||||||||||||||||||||
Literature Links: |
RNLS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNLS - renalase, FAD dependent amine oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031709.2 | 4901 | Intron | NP_001026879.2 | |||
NM_018363.3 | 4901 | Intron | NP_060833.1 | |||
XM_005269946.1 | 4901 | Intron | XP_005270003.1 | |||
XM_005269947.1 | 4901 | Intron | XP_005270004.1 | |||
XM_005269948.2 | 4901 | Intron | XP_005270005.1 | |||
XM_005269949.4 | 4901 | Intron | XP_005270006.1 | |||
XM_005269950.3 | 4901 | Intron | XP_005270007.1 | |||
XM_011539924.2 | 4901 | UTR 3 | XP_011538226.1 | |||
XM_011539927.2 | 4901 | Intron | XP_011538229.1 | |||
XM_017016380.1 | 4901 | Intron | XP_016871869.1 | |||
XM_017016381.1 | 4901 | Intron | XP_016871870.1 | |||
XM_017016382.1 | 4901 | UTR 3 | XP_016871871.1 | |||
XM_017016383.1 | 4901 | Intron | XP_016871872.1 | |||
XM_017016384.1 | 4901 | Intron | XP_016871873.1 | |||
XM_017016385.1 | 4901 | Intron | XP_016871874.1 |