Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BARD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BARD1 - BRCA1 associated RING domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000465.3 | 2031 | UTR 3 | NP_000456.2 | |||
NM_001282543.1 | 2031 | UTR 3 | NP_001269472.1 | |||
NM_001282545.1 | 2031 | UTR 3 | NP_001269474.1 | |||
NM_001282548.1 | 2031 | UTR 3 | NP_001269477.1 | |||
NM_001282549.1 | 2031 | UTR 3 | NP_001269478.1 | |||
XM_017004613.1 | 2031 | UTR 3 | XP_016860102.1 | |||
XM_017004614.1 | 2031 | Intron | XP_016860103.1 |