Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615295 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHSA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AHSA2 - AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321300.1 | Intron | NP_001308229.1 | ||||
NM_001321301.1 | Intron | NP_001308230.1 | ||||
NM_001321302.1 | Intron | NP_001308231.1 | ||||
NM_001321303.1 | Intron | NP_001308232.1 | ||||
NM_001321304.1 | Intron | NP_001308233.1 | ||||
NM_001321305.1 | Intron | NP_001308234.1 | ||||
NM_152392.4 | Intron | NP_689605.1 | ||||
XM_011532541.2 | Intron | XP_011530843.1 | ||||
XM_011532543.2 | Intron | XP_011530845.1 | ||||
XM_011532545.2 | Intron | XP_011530847.1 | ||||
XM_017003377.1 | Intron | XP_016858866.1 | ||||
XM_017003378.1 | Intron | XP_016858867.1 |
USP34 - ubiquitin specific peptidase 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |