Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603599 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFLAR PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFLAR - CASP8 and FADD like apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127183.2 | Intron | NP_001120655.1 | ||||
NM_001127184.2 | Intron | NP_001120656.1 | ||||
NM_001202515.1 | Intron | NP_001189444.1 | ||||
NM_001202516.1 | Intron | NP_001189445.1 | ||||
NM_001202517.1 | Intron | NP_001189446.1 | ||||
NM_001202518.1 | Intron | NP_001189447.1 | ||||
NM_001202519.1 | Intron | NP_001189448.1 | ||||
NM_001308042.1 | Intron | NP_001294971.1 | ||||
NM_001308043.1 | Intron | NP_001294972.1 | ||||
NM_003879.5 | Intron | NP_003870.4 | ||||
XM_017005189.1 | Intron | XP_016860678.1 | ||||
XM_017005190.1 | Intron | XP_016860679.1 | ||||
XM_017005191.1 | Intron | XP_016860680.1 | ||||
XM_017005192.1 | Intron | XP_016860681.1 | ||||
XM_017005193.1 | Intron | XP_016860682.1 | ||||
XM_017005194.1 | Intron | XP_016860683.1 | ||||
XM_017005195.1 | Intron | XP_016860684.1 | ||||
XM_017005196.1 | Intron | XP_016860685.1 | ||||
XM_017005197.1 | Intron | XP_016860686.1 | ||||
XM_017005198.1 | Intron | XP_016860687.1 |
Set Membership: |
HapMap |