Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608155 | ||||||||||||||||||||
Literature Links: |
SYNPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYNPO - synaptopodin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109974.2 | Intron | NP_001103444.1 | ||||
NM_001166208.1 | Intron | NP_001159680.1 | ||||
NM_001166209.1 | Intron | NP_001159681.1 | ||||
NM_007286.5 | Intron | NP_009217.3 | ||||
XM_005268370.1 | Intron | XP_005268427.1 | ||||
XM_005268371.1 | Intron | XP_005268428.1 | ||||
XM_006714755.3 | Intron | XP_006714818.1 | ||||
XM_011537552.2 | Intron | XP_011535854.1 | ||||
XM_017009007.1 | Intron | XP_016864496.1 | ||||
XM_017009008.1 | Intron | XP_016864497.1 |