Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616904 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOCK5 - dedicator of cytokinesis 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322810.1 | Intron | NP_001309739.1 | ||||
NM_024940.7 | Intron | NP_079216.4 | ||||
XM_011544661.1 | Intron | XP_011542963.1 | ||||
XM_011544662.1 | Intron | XP_011542964.1 | ||||
XM_011544663.2 | Intron | XP_011542965.1 | ||||
XM_017013865.1 | Intron | XP_016869354.1 | ||||
XM_017013866.1 | Intron | XP_016869355.1 | ||||
XM_017013867.1 | Intron | XP_016869356.1 | ||||
XM_017013868.1 | Intron | XP_016869357.1 |
Set Membership: |
HapMap |