Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612849 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP46 - ubiquitin specific peptidase 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134223.1 | 2037 | UTR 3 | NP_001127695.1 | |||
NM_001286767.1 | 2037 | UTR 3 | NP_001273696.1 | |||
NM_001286768.1 | 2037 | UTR 3 | NP_001273697.1 | |||
NM_022832.3 | 2037 | UTR 3 | NP_073743.2 | |||
XM_017008557.1 | 2037 | UTR 3 | XP_016864046.1 |
Set Membership: |
HapMap |