Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605434 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLSPN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLSPN - claspin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190481.1 | 5231 | UTR 3 | NP_001177410.1 | |||
NM_022111.3 | 5231 | UTR 3 | NP_071394.2 | |||
XM_011541938.2 | 5231 | UTR 3 | XP_011540240.1 | |||
XM_011541939.2 | 5231 | Intron | XP_011540241.1 | |||
XM_011541940.2 | 5231 | Intron | XP_011540242.1 | |||
XM_011541941.2 | 5231 | UTR 3 | XP_011540243.1 |