Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300415 | ||||||||||||||||||||
Literature Links: |
MTM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTM1 - myotubularin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000252.2 | Intron | NP_000243.1 | ||||
XM_005274687.2 | Intron | XP_005274744.1 | ||||
XM_011531171.1 | Intron | XP_011529473.1 | ||||
XM_011531172.1 | Intron | XP_011529474.1 | ||||
XM_011531173.2 | Intron | XP_011529475.1 | ||||
XM_017029547.1 | Intron | XP_016885036.1 | ||||
XM_017029548.1 | Intron | XP_016885037.1 | ||||
XM_017029549.1 | Intron | XP_016885038.1 | ||||
XM_017029550.1 | Intron | XP_016885039.1 | ||||
XM_017029551.1 | Intron | XP_016885040.1 |