Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120900 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C5 - complement component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317163.1 | Intron | NP_001304092.1 | ||||
NM_001317164.1 | Intron | NP_001304093.1 | ||||
NM_001735.2 | Intron | NP_001726.2 | ||||
XM_011518980.2 | Intron | XP_011517282.1 | ||||
XM_017015102.1 | Intron | XP_016870591.1 | ||||
XM_017015103.1 | Intron | XP_016870592.1 | ||||
XM_017015104.1 | Intron | XP_016870593.1 |
Set Membership: |
HapMap |