Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600041 | ||||||||||||||||||||
Literature Links: |
P2RY2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
P2RY2 - purinergic receptor P2Y2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002564.3 | Intron | NP_002555.3 | ||||
NM_176071.2 | Intron | NP_788085.2 | ||||
NM_176072.2 | Intron | NP_788086.2 | ||||
XM_005274019.4 | Intron | XP_005274076.1 | ||||
XM_005274020.4 | Intron | XP_005274077.1 | ||||
XM_005274021.4 | Intron | XP_005274078.1 | ||||
XM_011545074.2 | Intron | XP_011543376.1 | ||||
XM_017017839.1 | Intron | XP_016873328.1 |