Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601595 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMAD1 - SMAD family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003688.1 | Intron | NP_001003688.1 | ||||
NM_005900.2 | Intron | NP_005891.1 | ||||
XM_005262992.3 | Intron | XP_005263049.1 | ||||
XM_006714217.2 | Intron | XP_006714280.1 | ||||
XM_011531961.1 | Intron | XP_011530263.1 | ||||
XM_011531962.1 | Intron | XP_011530264.1 | ||||
XM_011531963.1 | Intron | XP_011530265.1 | ||||
XM_011531964.1 | Intron | XP_011530266.1 |
Set Membership: |
HapMap |