Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606738 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OSBPL10 - oxysterol binding protein like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174060.1 | Intron | NP_001167531.1 | ||||
NM_017784.4 | Intron | NP_060254.2 | ||||
XM_005264843.3 | Intron | XP_005264900.1 | ||||
XM_005264844.4 | Intron | XP_005264901.1 | ||||
XM_005264845.1 | Intron | XP_005264902.1 | ||||
XM_011533326.2 | Intron | XP_011531628.1 | ||||
XM_017005667.1 | Intron | XP_016861156.1 | ||||
XM_017005668.1 | Intron | XP_016861157.1 | ||||
XM_017005669.1 | Intron | XP_016861158.1 | ||||
XM_017005670.1 | Intron | XP_016861159.1 |
ZNF860 - zinc finger protein 860 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137674.2 | Intron | NP_001131146.2 | ||||
XM_017006299.1 | Intron | XP_016861788.1 |