Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATGAAAAGTTACGTGCTACTA[C/G]GATACTGAGTTTAGTTCCTAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP1A - FK506 binding protein 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000801.4 | Intron | NP_000792.1 | ||||
NM_001199786.1 | Intron | NP_001186715.1 | ||||
NM_054014.3 | Intron | NP_463460.1 |
FKBP1A-SDCBP2 - FKBP1A-SDCBP2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDCBP2-AS1 - SDCBP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |