Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603347 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPAS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPAS2 - neuronal PAS domain protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002518.3 | Intron | NP_002509.2 | ||||
XM_005263953.2 | Intron | XP_005264010.1 | ||||
XM_005263959.2 | Intron | XP_005264016.1 | ||||
XM_005263960.2 | Intron | XP_005264017.1 | ||||
XM_005263961.4 | Intron | XP_005264018.1 | ||||
XM_011511242.2 | Intron | XP_011509544.1 | ||||
XM_011511243.2 | Intron | XP_011509545.1 | ||||
XM_017004214.1 | Intron | XP_016859703.1 | ||||
XM_017004215.1 | Intron | XP_016859704.1 | ||||
XM_017004216.1 | Intron | XP_016859705.1 | ||||
XM_017004217.1 | Intron | XP_016859706.1 |