Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616014 MIM: 607652 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF25 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF25 - ring finger protein 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK36 - serine/threonine kinase 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243313.1 | Intron | NP_001230242.1 | ||||
NM_015690.4 | Intron | NP_056505.2 | ||||
XM_005246464.1 | Intron | XP_005246521.1 | ||||
XM_011510959.1 | Intron | XP_011509261.1 | ||||
XM_011510960.1 | Intron | XP_011509262.1 | ||||
XM_011510961.1 | Intron | XP_011509263.1 | ||||
XM_017003804.1 | Intron | XP_016859293.1 | ||||
XM_017003805.1 | Intron | XP_016859294.1 |
Set Membership: |
HapMap |