Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614956 MIM: 614958 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLFN12L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLFN12L - schlafen family member 12 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLFN14 - schlafen family member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129820.1 | 4345 | Intron | NP_001123292.1 | |||
XM_017024576.1 | 4345 | UTR 3 | XP_016880065.1 | |||
XM_017024577.1 | 4345 | UTR 3 | XP_016880066.1 | |||
XM_017024578.1 | 4345 | UTR 3 | XP_016880067.1 | |||
XM_017024579.1 | 4345 | UTR 3 | XP_016880068.1 |