Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602626 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL1RAP - interleukin 1 receptor accessory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167928.1 | Intron | NP_001161400.1 | ||||
NM_001167929.1 | Intron | NP_001161401.1 | ||||
NM_001167930.1 | Intron | NP_001161402.1 | ||||
NM_001167931.1 | Intron | NP_001161403.1 | ||||
NM_002182.3 | Intron | NP_002173.1 | ||||
NM_134470.3 | Intron | NP_608273.1 | ||||
XM_017006347.1 | Intron | XP_016861836.1 | ||||
XM_017006348.1 | Intron | XP_016861837.1 |
Set Membership: |
HapMap |