Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182134 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR1F PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HTR1F - 5-hydroxytryptamine receptor 1F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000866.4 | Intron | NP_000857.1 | ||||
NM_001322208.1 | Intron | NP_001309137.1 | ||||
NM_001322209.1 | Intron | NP_001309138.1 | ||||
NM_001322210.1 | Intron | NP_001309139.1 | ||||
XM_005264751.3 | Intron | XP_005264808.1 | ||||
XM_011533664.2 | Intron | XP_011531966.1 |
Set Membership: |
HapMap |