Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACCAGGCATGATAGATACAATT[A/C]TAATGAAGTCTTTGCAATGACCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603477 MIM: 607075 MIM: 614774 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5J2-PTCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP5J2-PTCD1 - ATP5J2-PTCD1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BUD31 - BUD31 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003910.3 | Intron | NP_003901.2 | ||||
XM_005250670.4 | Intron | XP_005250727.1 | ||||
XM_005250671.4 | Intron | XP_005250728.1 | ||||
XM_005250674.3 | Intron | XP_005250731.1 | ||||
XM_017012760.1 | Intron | XP_016868249.1 | ||||
XM_017012761.1 | Intron | XP_016868250.1 | ||||
XM_017012762.1 | Intron | XP_016868251.1 |
PDAP1 - PDGFA associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCD1 - pentatricopeptide repeat domain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |