Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610881 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT5B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KMT5B - lysine methyltransferase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300907.1 | 3261 | UTR 3 | NP_001287836.1 | |||
NM_001300908.1 | 3261 | UTR 3 | NP_001287837.1 | |||
NM_001300909.1 | 3261 | Intron | NP_001287838.1 | |||
NM_016028.4 | 3261 | Intron | NP_057112.3 | |||
NM_017635.4 | 3261 | UTR 3 | NP_060105.3 | |||
XM_005274035.3 | 3261 | UTR 3 | XP_005274092.2 | |||
XM_005274036.3 | 3261 | UTR 3 | XP_005274093.2 | |||
XM_006718581.1 | 3261 | UTR 3 | XP_006718644.1 | |||
XM_011545091.1 | 3261 | UTR 3 | XP_011543393.1 | |||
XM_011545092.2 | 3261 | UTR 3 | XP_011543394.1 | |||
XM_011545093.2 | 3261 | UTR 3 | XP_011543395.1 | |||
XM_011545094.2 | 3261 | Intron | XP_011543396.1 | |||
XM_017017876.1 | 3261 | UTR 3 | XP_016873365.1 | |||
XM_017017877.1 | 3261 | UTR 3 | XP_016873366.1 | |||
XM_017017878.1 | 3261 | UTR 3 | XP_016873367.1 | |||
XM_017017879.1 | 3261 | UTR 3 | XP_016873368.1 | |||
XM_017017880.1 | 3261 | Intron | XP_016873369.1 |