Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600168 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MST1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MST1R - macrophage stimulating 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244937.2 | 4338 | Intron | NP_001231866.1 | |||
NM_001318913.1 | 4338 | Intron | NP_001305842.1 | |||
NM_002447.3 | 4338 | Intron | NP_002438.2 | |||
XM_005265170.4 | 4338 | Intron | XP_005265227.2 | |||
XM_011533739.2 | 4338 | Intron | XP_011532041.1 | |||
XM_011533740.2 | 4338 | Intron | XP_011532042.1 | |||
XM_011533741.2 | 4338 | Intron | XP_011532043.1 | |||
XM_011533742.2 | 4338 | Intron | XP_011532044.1 | |||
XM_011533743.2 | 4338 | UTR 3 | XP_011532045.1 | |||
XM_011533744.2 | 4338 | Intron | XP_011532046.1 |
Set Membership: |
HapMap |