Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPXM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CPXM2 - carboxypeptidase X (M14 family), member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198148.2 | Intron | NP_937791.2 | ||||
XM_005269528.3 | Intron | XP_005269585.1 | ||||
XM_011539283.2 | Intron | XP_011537585.1 | ||||
XM_011539285.2 | Intron | XP_011537587.1 | ||||
XM_011539286.1 | Intron | XP_011537588.1 | ||||
XM_017015673.1 | Intron | XP_016871162.1 | ||||
XM_017015674.1 | Intron | XP_016871163.1 |
Set Membership: |
HapMap |