Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAATTCTGAGCTTCATTCTGTA[C/T]CAAATAACTTCTCAAATTTCCGATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605214 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNMB2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNMB2 - potassium calcium-activated channel subfamily M regulatory beta subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278911.1 | Intron | NP_001265840.1 | ||||
NM_005832.4 | Intron | NP_005823.1 | ||||
NM_181361.2 | Intron | NP_852006.1 | ||||
XM_011512325.2 | Intron | XP_011510627.1 |
KCNMB2-AS1 - KCNMB2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |