Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607740 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf64 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C17orf64 - chromosome 17 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181707.2 | Intron | NP_859058.2 | ||||
XM_005257033.4 | Intron | XP_005257090.1 | ||||
XM_005257034.4 | Intron | XP_005257091.1 | ||||
XM_005257035.4 | Intron | XP_005257092.1 | ||||
XM_005257036.2 | Intron | XP_005257093.1 | ||||
XM_017024166.1 | Intron | XP_016879655.1 |
RPL12P38 - ribosomal protein L12 pseudogene 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP32 - ubiquitin specific peptidase 32 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |